missing translation for 'onlineSavingsMsg'
Learn More

1,2-Diiodoethane, 98%

Product Code. 11361996
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11361996 25 g 25g
11351996 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11361996 Supplier Thermo Scientific Alfa Aesar Supplier No. L03019.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 624-73-7 | C2H4I2 | 281.863 g/mol

1,2-Diiodoethane is used as C-X bond formation reagent. It is also used as Pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Diiodoethane is used as C-X bond formation reagent. It is also used as Pharmaceutical intermediates.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Store at 4°C. Air & Light Sensitive. Incompatible with strong bases, strong oxidizing agents, heat and light. Protect from heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 624-73-7
Molecular Formula C2H4I2
Molecular Weight (g/mol) 281.863
MDL Number MFCD00001093
InChI Key GBBZLMLLFVFKJM-UHFFFAOYSA-N
Synonym ethane, 1,2-diiodo, ethylene diiodide, ethylene iodide, 1,2-diiodethane, 1,2-diiodo ethane, 1,2-bis iodanyl ethane, ch2ich2i, 1,2-diiodoethane, acmc-1b2u1, ksc358c5b
PubChem CID 12224
IUPAC Name 1,2-diiodoethane
SMILES C(CI)I

Specifications

Melting Point 80°C to 84°C
Density 2.13
Quantity 25 g
Beilstein 1730870
Sensitivity Light sensitive
Solubility Information Soluble in methanol. Insoluble in water.
Formula Weight 281.86
Percent Purity 98%
Chemical Name or Material 1,2-Diiodoethane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.