missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dihydroxybenzene-3,5-disulfonic acid disodium salt monohydrate, 97%

Code produit 11400303
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Conditionnement:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantity unitSize
11400303 500 g 500g
11480293 25 g 25g
11490293 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 11400303 Fournisseur Thermo Scientific Alfa Aesar Code fournisseur A11439.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 270573-71-2 | C6H6Na2O9S2 | 332.205 g/mol

1,2-Dihydroxybenzene-3,5-disulfonic acid disodium salt monohydrate, is used as a colorimetric reagent for iron, manganese, titanium, molybdenum, and complexes of zirconium. It is used as a metal indicator. It is used for Chelate titration, colorimetry, fluorometry and masking reagent. It is used in chemical research, pharmaceutical research and as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Dihydroxybenzene-3,5-disulfonic acid disodium salt monohydrate, is used as a colorimetric reagent for iron, manganese, titanium, molybdenum, and complexes of zirconium. It is used as a metal indicator. It is used for Chelate titration, colorimetry, fluorometry and masking reagent. It is used in chemical research, pharmaceutical research and as a pharmaceutical intermediate.

Solubility
It is readily soluble in water.

Notes
Stable under normal conditions. Keep away from strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 270573-71-2
Molecular Formula C6H6Na2O9S2
Molecular Weight (g/mol) 332.205
MDL Number MFCD00007473
InChI Key ZLRROLLKQDRDPI-UHFFFAOYSA-L
Synonym tiron monohydrate, sodium 4,5-dihydroxybenzene-1,3-disulfonate hydrate, unii-kht84q5m4r, 4,5-dihydroxy-1,3-benzenedisulfonic acid disodium salt monohydrate, kht84q5m4r, disodium 4,5-dihydroxybenzene-1,3-disulfonate hydrate, 1,2-dihydroxybenzene-3,5-disulfonic acid disodium salt monohydrate, 1,3-benzenedisulfonicacid, 4,5-dihydroxy-, sodium salt, hydrate 1:2:1, 4,5-dihydroxybenzene-1,3-disulfonic acid, sodium salt, sodium salt, hydrate, sdd
PubChem CID 2723960
IUPAC Name disodium;4,5-dihydroxybenzene-1,3-disulfonate;hydrate
SMILES C1=C(C=C(C(=C1S(=O)(=O)[O-])O)O)S(=O)(=O)[O-].O.[Na+].[Na+]

Spécification

Melting Point >300°C
Quantity 500 g
Beilstein 4629740
Merck Index 14,9465
Solubility Information It is readily soluble in water.
Formula Weight 332.23 (314.21 Anhydrous)
Percent Purity 97%
Chemical Name or Material 1,2-Dihydroxybenzene-3,5-disulfonic acid disodium salt monohydrate

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.