missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dihydro Prednicarbate, TRC

Product Code. 30382861
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382861 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30382861 Supplier Toronto Research Chemicals Supplier No. TRCD44643525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2-dihydroprednicarbate
CAS 671225-26-6
Molecular Formula C27 H38 O8
Quantity 25 mg
Synonym Pregn-4-ene-3,20-dione, 17-[(ethoxycarbonyl)oxy]-11-hydroxy-21-(1-oxopropoxy)-, (11β)- (9CI), Cortisol 17-(ethyl carbonate) 21-propionate, 1,2-Dihydroprednicarbate, 1,2-Dihydro prednicarbate
InChI Formula InChI=1S/C27H38O8/c1-5-22(31)34-15-21(30)27(35-24(32)33-6-2)12-10-19-18-8-7-16-13-17(28)9-11-25(16,3)23(18)20(29)14-26(19,27)4/h13,18-20,23,29H,5-12,14-15H2,1-4H3/t18-,19-,20-,23+,25-,26-,27-/m0/s1
SMILES CCOC(=O)O[C@@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3[C@@H](O)C[C@]12C)C(=O)COC(=O)CC
IUPAC Name [2-[(8S,9S,10R,11S,13S,14S,17R)-17-ethoxycarbonyloxy-11-hydroxy-10,13-dimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl]-2-oxoethyl] propanoate
Molecular Weight (g/mol) 490.59
Formula Weight 490.2567
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.