missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dihydro-oseltamivir (mixture of diastereomers), TRC

Product Code. 30387402
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387402 50 mg -
30374832 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30387402 Supplier Toronto Research Chemicals Supplier No. TRCD45065850MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1052063-38-3
Molecular Formula C16 H30 N2 O4
Molecular Weight (g/mol) 314.42
Synonym (3S,4R,5R)-4-(Acetylamino)-3-amino-5-(1-ethylpropoxy)-cyclohexanecarboxylic Acid Ethyl Ester
IUPAC Name ethyl (3S,4R,5R)-4-acetamido-3-amino-5-(1-ethylpropoxy)cyclohexanecarboxylate
SMILES CCOC(=O)C1C[C@H](N)[C@@H](NC(=O)C)[C@@H](C1)OC(CC)CC

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C16H30N2O4/c1-5-12(6-2)22-14-9-11(16(20)21-7-3)8-13(17)15(14)18-10(4)19/h11-15H,5-9,17H2,1-4H3,(H,18,19)/t11?,13-,14+,15+/m0/s1
Formula Weight 314.2206
Chemical Name or Material 1,2-Dihydro Oseltamivir
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.