missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Dihydro Betamethasone 9,11-Epoxide 17,21-Dipropionate, TRC
Specifications
Specifications
| Chemical Name or Material | 1,2-Dihydro Betamethasone 9,11-Epoxide 17,21-Dipropionate |
| Analyte or Component Names | 1,2-Dihydro Betamethasone 9,11-Epoxide 17,21-Dipropionate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C28 H38 O7 |
| Synonym | 9beta,11beta,16beta)-9,11-Epoxy-16-methyl-17,21-bis(1-oxopropoxy)pregna-4-ene-3,20-dione |
| InChI Formula | InChI=1S/C28H38O7/c1-6-23(31)33-15-21(30)27(35-24(32)7-2)16(3)12-20-19-9-8-17-13-18(29)10-11-25(17,4)28(19)22(34-28)14-26(20,27)5/h13,16,19-20,22H,6-12,14-15H2,1-5H3/t16-,19-,20-,22-,25-,26-,27-,28+/m0/s1 |
| Solution Type | Neat |
| SMILES | CCC(=O)OCC(=O)[C@@]1(OC(=O)CC)[C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@@]35O[C@H]5C[C@]12C |
| IUPAC Name | [2-oxo-2-[(1S,2S,10S,11S,13S,14R,15S,17S)-2,13,15-trimethyl-5-oxo-14-propanoyloxy-18-oxapentacyclo[8.8.0.01,17.02,7.011,15]octadec-6-en-14-yl]ethyl] propanoate |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?