missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dihydro Betamethasone 9,11-Epoxide 17,21-Dipropionate, TRC

Product Code. 30381403
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381403 5 mg 5mg
30384731 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30381403 Supplier Toronto Research Chemicals Supplier No. TRCD4486305MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2-Dihydro Betamethasone 9,11-Epoxide 17,21-Dipropionate
Analyte or Component Names 1,2-Dihydro Betamethasone 9,11-Epoxide 17,21-Dipropionate
Shipping Condition Room Temperature
Molecular Formula C28 H38 O7
Synonym 9beta,11beta,16beta)-9,11-Epoxy-16-methyl-17,21-bis(1-oxopropoxy)pregna-4-ene-3,20-dione
InChI Formula InChI=1S/C28H38O7/c1-6-23(31)33-15-21(30)27(35-24(32)7-2)16(3)12-20-19-9-8-17-13-18(29)10-11-25(17,4)28(19)22(34-28)14-26(20,27)5/h13,16,19-20,22H,6-12,14-15H2,1-5H3/t16-,19-,20-,22-,25-,26-,27-,28+/m0/s1
Solution Type Neat
SMILES CCC(=O)OCC(=O)[C@@]1(OC(=O)CC)[C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@@]35O[C@H]5C[C@]12C
IUPAC Name [2-oxo-2-[(1S,2S,10S,11S,13S,14R,15S,17S)-2,13,15-trimethyl-5-oxo-14-propanoyloxy-18-oxapentacyclo[8.8.0.01,17.02,7.011,15]octadec-6-en-14-yl]ethyl] propanoate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 486.597
Quantity 5 mg
Formula Weight 486.262
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.