missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dihydro Beclomethasone Dipropionate, TRC

Product Code. 30384209
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384209 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30384209 Supplier Toronto Research Chemicals Supplier No. TRCD448400100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2-Dihydro Beclomethasone Dipropionate
CAS 114371-33-4
Molecular Formula C28H39ClO7
Quantity 100 mg
Synonym (11β,16β)-9-chloro-11-hydroxy-16-methyl-17,21-bis(1-oxopropoxy)pregn-4-ene-3,20-dione, 1,2-Dihydrobeclometasone Dipropopionate, Beclomethasone Dipropionate EP Impurity L
InChI Formula InChI=1S/C28H39ClO7/c1-6-23(33)35-15-22(32)28(36-24(34)7-2)16(3)12-20-19-9-8-17-13-18(30)10-11-25(17,4)27(19,29)21(31)14-26(20,28)5/h13,16,19-21,31H,6-12,14-15H2,1-5H3/t16-,19-,20-,21-,25-,26-,27-,28-/m0/s1
SMILES CCC(=O)OCC(=O)[C@]1([C@H](C[C@@H]2[C@@]1(C[C@@H]([C@]3([C@H]2CCC4=CC(=O)CC[C@@]43C)Cl)O)C)C)OC(=O)CC
Molecular Weight (g/mol) 523.06
Formula Weight 522.2384
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.