missing translation for 'onlineSavingsMsg'
Learn More

1,2-Diethylbenzene, 97%

Product Code. 11379515
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11379515 50 g 50g
11359515 2 g 2g
11369515 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11379515 Supplier Thermo Scientific Alfa Aesar Supplier No. L18775.18
 Restricted Item
Estimated Shipment: 17-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 135-01-3 | C10H14 | 134.222 g/mol

1,2-Diethylbenzene is used as an intermediate and a building block in organic synthesis. 1,2-Diethylbenzene is used as one of the chemical composition of essential oils from flower and leaf of korean mint.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Diethylbenzene is used as an intermediate and a building block in organic synthesis. 1,2-Diethylbenzene is used as one of the chemical composition of essential oils from flower and leaf of korean mint.

Solubility
Miscible with alcohol, benzene and carbon tetrachloride. Immiscible with water.

Notes
Keep away from sources of ignition. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135-01-3
Molecular Formula C10H14
Molecular Weight (g/mol) 134.222
MDL Number MFCD00009258
InChI Key KVNYFPKFSJIPBJ-UHFFFAOYSA-N
Synonym o-diethylbenzene, benzene, 1,2-diethyl, benzene, diethyl, benzene, o-diethyl, diethylbenzol, unii-25hox6t1lu, 25hox6t1lu, diethylbenzenes mixed isomer, dsstox_cid_7866, dsstox_rid_78595
PubChem CID 8657
IUPAC Name 1,2-diethylbenzene
SMILES CCC1=CC=CC=C1CC

Specifications

Melting Point -31°C
Density 0.88
Boiling Point 183°C
Flash Point 55°C (131°F)
Refractive Index 1.505
Quantity 50 g
UN Number UN2049
Beilstein 1904392
Solubility Information Miscible with alcohol,benzene and carbon tetrachloride. Immiscible with water.
Formula Weight 134.22
Percent Purity 97%
Chemical Name or Material 1,2-Diethylbenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.