missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dicyanobenzene, 98%

Product Code. 10489470 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10489470 1 kg Plastic bottle 1kg
10255990 250 g Plastic bottle 250g
10337500 5 kg Glass bottle 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10489470 Supplier Thermo Scientific Acros Supplier No. 174010010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 91-15-6 | C8H4N2 | 128.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-15-6
Molecular Formula C8H4N2
Molecular Weight (g/mol) 128.13
MDL Number MFCD00001771
InChI Key XQZYPMVTSDWCCE-UHFFFAOYSA-N
Synonym phthalonitrile, 1,2-dicyanobenzene, o-phthalodinitrile, phthalodinitrile, 1,2-benzenedicarbonitrile, o-dicyanobenzene, 1,2-benzodinitrile, o-pdn, o-cyanobenzonitrile, o-benzenedicarbonitrile
PubChem CID 7042
IUPAC Name benzene-1,2-dicarbonitrile
SMILES N#CC1=CC=CC=C1C#N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 138.0°C to 142.0°C
Color White to Beige
Density 1.2400g/mL
Flash Point >130°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic bottle
Linear Formula C6H4(CN)2
Quantity 1 kg
Beilstein 09, 815
Specific Gravity 1.24
Solubility Information Solubility in water: 0.56g/L (25°C). Other solubilities: soluble in acetone and benzene,soluble in ether and alcohol
Formula Weight 128.13
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 1, 2-Dicyanobenzene, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.