missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichloropropane, 98%, pure

Product Code. 10274280 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
50 mL
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
50mL
5mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10274280 50 mL Glass bottle 50mL
10162520 5 mL Glass bottle 5mL
10137903 250 mL Glass bottle 250mL
10740975 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10274280 Supplier Thermo Scientific Acros Supplier No. 113670500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 78-87-5 | C3H6Cl2 | 112.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-87-5
Molecular Formula C3H6Cl2
Molecular Weight (g/mol) 112.98
MDL Number MFCD00000868
InChI Key KNKRKFALVUDBJE-UHFFFAOYNA-N
Synonym propylene dichloride, propane, 1,2-dichloro, propylene chloride, dwuchloropropan, bichlorure de propylene, rcra waste number u083, caswell no. 324, dwuchloropropan polish, alpha,beta-dichloropropane, ccris 951
PubChem CID 6564
ChEBI CHEBI:82163
IUPAC Name 1,2-dichloropropane
SMILES CC(Cl)CCl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -100.0°C
Color Colorless to Yellow
Density 1.1560g/mL
Boiling Point 95.0°C to 96.0°C
Flash Point 15°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH(Cl)CH2Cl
Refractive Index 1.4380 to 1.4400
Quantity 50 mL
Beilstein 01, 105
Merck Index 15, 7967
Specific Gravity 1.156
Solubility Information Solubility in water: 3g/L (20°C). Other solubilities: soluble in most common organic solvents
Formula Weight 112.99
Percent Purity 98%
Grade Pure
Physical Form Liquid
Chemical Name or Material 1, 2-Dichloropropane, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.