missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1,2-Dichlorohexafluorocyclopentene, 97%

Product Code. 11440177
Change view
Click to view available options
Quantity:
5g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11440177 5g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11440177 Supplier Alfa Aesar™ Supplier No. B21630.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 706-79-6
Boiling Point 90°C
Molecular Formula C5Cl2F6
MDL Number MFCD00019302
Quantity 5g
UN Number UN2810
Beilstein 2053012
Synonym 1,2-dichlorohexafluorocyclopentene, 1,2-dichlorohexafluoro-1-cyclopentene, cyclopentene-26, 1,2-dichlorohexafluorocyclopent-1-ene, 1,2-dichloroperfluorocyclopentene, cyclopentene, 1,2-dichloro-3,3,4,4,5,5-hexafluoro, cyclopentene, 1,2-dichlorohexafluoro, 1,2-dichlorohexafluorocyclopentene-1, 1,2-dichloro-3,3,4,4,5,5-hexafluorocyclopent-1-ene, hexafluoro-1,2-dichloro-1-cyclopentene
InChI Key ABPBVCKGWWGZDP-UHFFFAOYSA-N
SMILES C1(=C(C(C(C1(F)F)(F)F)(F)F)Cl)Cl
IUPAC Name 1,2-dichloro-3,3,4,4,5,5-hexafluorocyclopentene
Molecular Weight (g/mol) 244.945
PubChem CID 12815
Formula Weight 244.95
Percent Purity 97%
Chemical Name or Material 1,2-Dichlorohexafluorocyclopentene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.