missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichloroethanol Acetate, TRC

Product Code. 30380606
Change view
Click to view available options
Quantity:
250 mg
1 g
2.5 g
Unit Size:
1g
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380606 250 mg -
30386706 1 g 1g
30386962 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30380606 Supplier Toronto Research Chemicals Supplier No. TRCD434615250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10140-87-1
Molecular Formula C4H6Cl2O2
Molecular Weight (g/mol) 157
Synonym 1,2-Dichloroethyl Acetate
IUPAC Name 1,2-dichloroethyl acetate
SMILES CC(=O)OC(Cl)CCl

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C4H6Cl2O2/c1-3(7)8-4(6)2-5/h4H,2H2,1H3
Formula Weight 155.9745
Chemical Name or Material 1,2-Dichloroethanol Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.