missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichloroethane, HPLC Grade, 99% min

Product Code. 11338187
Change view
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
Unit Size:
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11338187 4 L 4 L
11318187 1 L 1 L
11328187 4 x 1 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11338187 Supplier Thermo Scientific Alfa Aesar Supplier No. 022918.K7
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 107-06-2 | C2H4Cl2 | 98.95 g/mol

1,2-Dichloroethane is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-06-2
Molecular Formula C2H4Cl2
Molecular Weight (g/mol) 98.95
MDL Number MFCD00000963
InChI Key WSLDOOZREJYCGB-UHFFFAOYSA-N
Synonym ethylene dichloride, ethylene chloride, ethane, 1,2-dichloro, dutch liquid, dutch oil, ethane dichloride, glycol dichloride, aethylenchlorid, dichloro-1,2-ethane, dichloremulsion
PubChem CID 11
ChEBI CHEBI:27789
IUPAC Name 1,2-dichloroethane
SMILES ClCCCl

Specifications

Melting Point -35°C
Density 1.256 g/mL
Quantity 4 L
Flash Point 15°C (59°F)
Physical Form Liquid
Refractive Index 1.4448
UN Number UN1184
Beilstein 605264
Merck Index 14,3797
Boiling Point 83°C
Solubility Information Miscible with methanol,diethyl ether,n-octanol,acetone. Very slightly miscible with water.
Formula Weight 98.96
Percent Purity ≤99%
Grade HPLC
Chemical Name or Material 1,2-Dichloroethane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.