missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichloroethane-D4, TRC

Product Code. 30369416
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Product Code. Quantity unitSize
30369416 500 mg 500mg
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30369416

Brand: Toronto Research Chemicals TRCD434327500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2-Dichloroethane D4
CAS 17060-07-0
Molecular Formula C2 2H4 Cl2
Quantity 500 mg
Synonym Ethane-1,1,2,2-d4, dichloro- (7CI), 1,2-Dichloroethane-d4
InChI Formula InChI=1S/C2H4Cl2/c3-1-2-4/h1-2H2/i1D2,2D2
SMILES [2H]C([2H])(Cl)C([2H])([2H])Cl
IUPAC Name 1,2-dichloro-1,1,2,2-tetradeuterioethane
Molecular Weight (g/mol) 102.98
Formula Weight 101.9941
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.