missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichloroethane-D4, TRC

Product Code. 30372595
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity unitSize
30372595 1 g 1g
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30372595

Brand: Toronto Research Chemicals TRCD4343271G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2-Dichloroethane D4
CAS 17060-07-0
Molecular Formula C2 2H4 Cl2
Quantity 1 g
Synonym Ethane-1,1,2,2-d4, dichloro- (7CI), 1,2-Dichloroethane-d4
InChI Formula InChI=1S/C2H4Cl2/c3-1-2-4/h1-2H2/i1D2,2D2
SMILES [2H]C([2H])(Cl)C([2H])([2H])Cl
IUPAC Name 1,2-dichloro-1,1,2,2-tetradeuterioethane
Molecular Weight (g/mol) 102.98
Formula Weight 101.9941
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.