missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Dichloroethane D4 & Toluene D8 Mixture 528 1000 μg/mL in Methanol, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | 1,2-Dichloroethane D4∣Toluene D8 |
| CAS | 17060-07-0, 2037-26-5 |
| Solvent or Matrix | Methanol |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | 1,2-Dichloroethane D4 1000 μg/mL∣Toluene D8 1000 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?