missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichloroethane-d4 (Reagent grade), CDN

Product Code. 30323635
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
30323635 5 g 5g
30315831 1 g 1g
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30323635

Brand: CDN CDND01035G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2-Dichloroethane D4
CAS 17060-07-0
Molecular Formula C2 2H4 Cl2
Quantity 5 g
Synonym 1,2-Dichloroethane D4, Ethane-1,1,2,2-d4, 1,2-dichloro- (8 CI,9 CI), Ethane-1,1,2,2-d4, dichloro- (7 CI), 1,2-Dichloroethane-d4
InChI Formula InChI=1 S/C2H4Cl2/c3-1-2-4/h1-2H2/i1D2,2D2
SMILES [2 H]C([2 H])(Cl)C([2 H])([2 H])Cl
IUPAC Name 1,2-dichloro-1,1,2,2-tetradeuterioethane
Molecular Weight (g/mol) 102.98
Formula Weight 101.9941 g/mol
Percent Purity 99 atom % D, min 99% Chemical Purity
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.