missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Dichloroethane-d4 (Reagent grade), CDN
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Specifications
Specifications
| Chemical Name or Material | 1,2-Dichloroethane D4 |
| CAS | 17060-07-0 |
| Molecular Formula | C2 2H4 Cl2 |
| Quantity | 1 g |
| Synonym | 1,2-Dichloroethane D4, Ethane-1,1,2,2-d4, 1,2-dichloro- (8 CI,9 CI), Ethane-1,1,2,2-d4, dichloro- (7 CI), 1,2-Dichloroethane-d4 |
| InChI Formula | InChI=1 S/C2H4Cl2/c3-1-2-4/h1-2H2/i1D2,2D2 |
| SMILES | [2 H]C([2 H])(Cl)C([2 H])([2 H])Cl |
| IUPAC Name | 1,2-dichloro-1,1,2,2-tetradeuterioethane |
| Molecular Weight (g/mol) | 102.98 |
| Formula Weight | 101.9941 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction