missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichloroethane-d{4}, 99% (Isotopic), Thermo Scientific Chemicals

Catalog Number 11369967
Click to view available options
Quantity:
1 g
5 g
5 x 1 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
17060-07-0
C2H4Cl2
102.978
MFCD00037531
WSLDOOZREJYCGB-LNLMKGTHSA-N
1,2-dichloroethane-d4, 1,2-dichloroethane d4, ethylene-d4 dichloride, dichloro 2 h? ethane, 1,2-dichloroethane-d4, 99 atom % d, 1,2-dichloroethane-d4 98.0 atom % d, 1,2-dichloroethane-d4, analytical standard, 1,2-dichloroethane d4 2000 ng/microl in methanol
12197860
1,2-dichloro-1,1,2,2-tetradeuterioethane
C(CCl)Cl
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
17060-07-0
C2H4Cl2
102.978
MFCD00037531
WSLDOOZREJYCGB-LNLMKGTHSA-N
1,2-dichloroethane-d4, 1,2-dichloroethane d4, ethylene-d4 dichloride, dichloro 2 h? ethane, 1,2-dichloroethane-d4, 99 atom % d, 1,2-dichloroethane-d4 98.0 atom % d, 1,2-dichloroethane-d4, analytical standard, 1,2-dichloroethane d4 2000 ng/microl in methanol
12197860
1,2-dichloro-1,1,2,2-tetradeuterioethane
C(CCl)Cl

1,2-Dichloroethane-d4 is widely used as a solvent for NMR analysis, and in the synthesis of poly(ethylene-d4 oxide) as a chain termination reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Liquid
Density 1.307 g/mL
Flash Point 15°C (60°F)
Refractive Index 1.443
Quantity 1 g
Melting Point -35°C
Boiling Point 83°C
UN Number UN1184
Solubility Information Partially miscible with water.
Formula Weight 102.99
Percent Purity 99%
Chemical Name or Material 1,2-Dichloroethane-d4, Isotopic
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'