missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichloroethane, CHROMASOLV™, for HPLC, ≥99.8%, Honeywell Riedel-de Haen

Product Code. 15614650
Change view
Click to view available options
Quantity:
100 mL
1 L
Packaging:
Glass Bottle
Unit Size:
1 L
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15614650 100 mL Glass Bottle 100mL
15624650 1 L Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 15614650 Supplier Honeywell-Chromasolv Supplier No. 34872100ML

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CHROMASOLV™, for HPLC,≥99.8%

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-06-2
Molecular Formula C2H4Cl2
Molecular Weight (g/mol) 98.95
MDL Number MFCD00000963
InChI Key WSLDOOZREJYCGB-UHFFFAOYSA-N
Synonym ethylene dichloride, ethylene chloride, ethane, 1,2-dichloro, dutch liquid, dutch oil, ethane dichloride, glycol dichloride, aethylenchlorid, dichloro-1,2-ethane, dichloremulsion
PubChem CID 11
ChEBI CHEBI:27789
IUPAC Name 1,2-dichloroethane
SMILES ClCCCl

Specifications

Melting Point -35°C (literature)
Absorbance max. 0.01 at 270 nm, max. 0.04 at 245 nm, max. 0.60 at 230 nm
CAS Max % ≤100.0000%
Density 1.256 g/mL at 25 °C(lit.)
Quantity 100 mL
Flash Point 13°C
Packaging Glass Bottle
Linear Formula ClCH2CH2Cl
UN Number UN1184
Beilstein 605264
Boiling Point 83°C
Formula Weight 98.96
Percent Purity >99.8%
Grade HPLC
Chemical Name or Material 1,2-Dichloroethane, CHROMASOLV™, for HPLC, ≥99.8%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.