missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichloroethane, 99.5%, for electronic use (MOS), residue free

Product Code. 10357071 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10357071 1 L Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 10357071 Supplier Thermo Scientific Acros Supplier No. 327860010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 107-06-2 | C2H4Cl2 | 98.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For electronic use
TRUSTED_SUSTAINABILITY

Specifications

Melting Point -35.0°C
CAS 107-06-2
CAS Min % 99.4
Acidity 0.001% max.
CAS Max % 100.0
Density 1.2500g/mL
Quantity 1 L
Flash Point 13°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.4440 to 1.4460
Residue after Evaporation 0.0005% max.
Assay Percent Range 99.5%
Specific Gravity 1.25
Molecular Formula C2H4Cl2
Linear Formula ClCH2CH2Cl
MDL Number MFCD00000963
Viscosity 0.8 mPa.s (20°C)
Water 0.03% max. (Coulometric)
Beilstein 01, 84
Merck Index 15, 3851
Synonym ethylene dichloride, ethylene chloride, ethane, 1,2-dichloro, dutch liquid, dutch oil, ethane dichloride, glycol dichloride, aethylenchlorid, dichloro-1,2-ethane, dichloremulsion
Boiling Point 81.0°C to 85.0°C
Solubility Information Solubility in water: 8.7g/L (20°C). Other solubilities: miscible with most common organic solvents
InChI Key WSLDOOZREJYCGB-UHFFFAOYSA-N
SMILES ClCCCl
IUPAC Name 1,2-dichloroethane
Molecular Weight (g/mol) 98.95
PubChem CID 11
ChEBI CHEBI:27789
Formula Weight 98.96
Percent Purity 99.5%
Grade Electronic
Chemical Name or Material 1,2-Dichloroethane, Residue Free
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.