missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichloroethane, 99.5%, for analysis, Thermo Scientific Chemicals

Catalog Number p-4812564
28.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
107-06-2
C2H4Cl2
98.95
MFCD00000963
WSLDOOZREJYCGB-UHFFFAOYSA-N
1,2-dichloroethane
ClCCCl
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
107-06-2
C2H4Cl2
98.95
MFCD00000963
WSLDOOZREJYCGB-UHFFFAOYSA-N
1,2-dichloroethane
ClCCCl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -35.0°C
Density 1.2500g/mL
Boiling Point 81.0°C to 85.0°C
Quantity 1 L
Flash Point 13°C
Packaging Glass Bottle
Refractive Index 1.4440 to 1.4460
Assay Percent Range 99.4% min. (GC)
Specific Gravity 1.25
Linear Formula ClCH2CH2Cl
Viscosity 0.8 mPa.s (20°C)
Merck Index 15,3851
Solubility Information Solubility in water: 8.7g/L (20°C). Other solubilities: miscible with most common organic solvents
Formula Weight 98.96
Percent Purity 99.50%
Grade Analytical
Chemical Name or Material 1,2-Dichloroethane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'