missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichlorobenzene, TRC

Product Code. 30387305
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30387305 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30387305 Supplier Toronto Research Chemicals Supplier No. TRCD431870100G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 95-50-1
Molecular Formula C6 H4 Cl2
Quantity 100 g
Synonym Benzene, o-dichloro- (8CI), 1,2-Dichlorobenzene, Cloroben, Dilatin DB, Dowtherm E, NSC 60644, o-Dichlorobenzene
InChI Formula InChI=1S/C6H4Cl2/c7-5-3-1-2-4-6(5)8/h1-4H
SMILES Clc1ccccc1Cl
IUPAC Name 1,2-dichlorobenzene
Molecular Weight (g/mol) 147
Formula Weight 145.969
Chemical Name or Material 1,2-Dichlorobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.