missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichlorobenzene, HPLC Grade, 98% min, Thermo Scientific Chemicals

Catalog Number 11363547
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
This item is not returnable. View return policy
95-50-1
C6H4Cl2
146.998
MFCD00000535
RFFLAFLAYFXFSW-UHFFFAOYSA-N
o-dichlorobenzene, chloroben, o-dichlorbenzol, dilantin db, ortho-dichlorobenzene, 2-dichlorobenzene, o-dichlorobenzol, odcb, benzene, 1,2-dichloro, cloroben
7239
CHEBI:35290
1,2-dichlorobenzene
C1=CC=C(C(=C1)Cl)Cl
This item is not returnable. View return policy
95-50-1
C6H4Cl2
146.998
MFCD00000535
RFFLAFLAYFXFSW-UHFFFAOYSA-N
o-dichlorobenzene, chloroben, o-dichlorbenzol, dilantin db, ortho-dichlorobenzene, 2-dichlorobenzene, o-dichlorobenzol, odcb, benzene, 1,2-dichloro, cloroben
7239
CHEBI:35290
1,2-dichlorobenzene
C1=CC=C(C(=C1)Cl)Cl

1,2-Dichlorobenzene is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -18°C
Density 1.306 g/mL
Boiling Point 180°C
Flash Point 68°C (154°F)
Odor Pungent
Refractive Index 1.551
Quantity 1 L
UN Number UN1591
Beilstein 606078
Merck Index 14,3056
Solubility Information Miscible with water and most of the organic solvents.
Formula Weight 147
Percent Purity ≤98%
Grade HPLC
Physical Form Liquid
Chemical Name or Material 1,2-Dichlorobenzene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'