missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichlorobenzene, Honeywell™

Product Code. 15613920
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15613920 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15613920 Supplier Honeywell Riedel-de Haën Supplier No. 331601L

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 95-50-1
Density 1.306g/mL at 25°C
Explosion Limit 0.092
Flash Point 66°C (150.8°F)
Ignition Point 1198°F
Molecular Formula C6H4Cl2
MDL Number MFCD00000535
Quantity 1 L
UN Number UN1591
Beilstein 606078
Synonym o-dichlorobenzene, chloroben, o-dichlorbenzol, dilantin db, ortho-dichlorobenzene, 2-dichlorobenzene, o-dichlorobenzol, odcb, benzene, 1,2-dichloro, cloroben
InChI Key RFFLAFLAYFXFSW-UHFFFAOYSA-N
SMILES C1=CC=C(C(=C1)Cl)Cl
IUPAC Name 1,2-dichlorobenzene
Molecular Weight (g/mol) 146.998
PubChem CID 7239
Vapor Density 5.1 (vs air)
ChEBI CHEBI:35290
Formula Weight 147g/mol
Vapor Pressure 1.2 mmHg (20°C)1.6 mmHg (35°C)
Percent Purity ≥99% (GC)
Chemical Name or Material 1,2-Dichlorobenzene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.