missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichlorobenzene-d4, TRC

Product Code. 30378862
Change view
Click to view available options
Quantity:
10 mg
100 mg
Product Code. Quantity
30378862 100 mg
30371120 10 mg
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30378862

Brand: Toronto Research Chemicals TRCD431872100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2199-69-1
Formula Weight 149.9941
InChI Formula InChI=1S/C6H4Cl2/c7-5-3-1-2-4-6(5)8/h1-4H/i1D,2D,3D,4D
IUPAC Name 1,2-dichloro-3,4,5,6-tetradeuteriobenzene
Molecular Formula C6 2H4 Cl2
Molecular Weight (g/mol) 151.03
SMILES [2H]c1c([2H])c([2H])c(Cl)c(Cl)c1[2H]
Synonym Benzene-1,2,3,4-d4, dichloro- (7CI), 5,6-Dichlorobenzene-1,2,3,4-d4, 1,2-Dichlorobenzene-d4, 1,2-Dichlorotetradeuterobenzene, o-Dichlorobenzene-d4
Chemical Name or Material 1,2-Dichlorobenzene D4

Specifications

Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.