missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichlorobenzene-d{4}, 99% (Isotopic), Thermo Scientific Chemicals

Catalog Number 11359967
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
2199-69-1
C6H4Cl2
151.02
MFCD00037106
RFFLAFLAYFXFSW-RHQRLBAQSA-N
1,2-dichlorobenzene-d4, 1,2-dichlorobenzene d4, benzene-1,2,3,4-d4-, 5,6-dichloro, o-dichloro 2h4 benzene, deuterated 1,2-dichlorobenzene, tetradeutero-1,2-dichlorobenzene, 1,2-dichlorobenzene-d4, 98 atom % d, 1,2-dichlorobenzene-d4,98atom%d, benzene-1,2,3,4-d4, 5,6-dichloro, 1,2-dichloro-3,4,5,6-tetradeuterobenzene
519913
1,2-dichloro-3,4,5,6-tetradeuteriobenzene
[2H]C1=C([2H])C([2H])=C(Cl)C(Cl)=C1[2H]
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
2199-69-1
C6H4Cl2
151.02
MFCD00037106
RFFLAFLAYFXFSW-RHQRLBAQSA-N
1,2-dichlorobenzene-d4, 1,2-dichlorobenzene d4, benzene-1,2,3,4-d4-, 5,6-dichloro, o-dichloro 2h4 benzene, deuterated 1,2-dichlorobenzene, tetradeutero-1,2-dichlorobenzene, 1,2-dichlorobenzene-d4, 98 atom % d, 1,2-dichlorobenzene-d4,98atom%d, benzene-1,2,3,4-d4, 5,6-dichloro, 1,2-dichloro-3,4,5,6-tetradeuterobenzene
519913
1,2-dichloro-3,4,5,6-tetradeuteriobenzene
[2H]C1=C([2H])C([2H])=C(Cl)C(Cl)=C1[2H]

It is applied in NMR spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Liquid
Density 1.34 g/mL
Flash Point 65°C (150°F)
Refractive Index 1.5509
Quantity 5 g
Melting Point -17°C
Boiling Point 181°C
UN Number UN1591
Solubility Information Insoluble in water
Formula Weight 151.03
Percent Purity 99%
Chemical Name or Material 1,2-Dichlorobenzene-d4, Isotopic
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'