missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichlorobenzene, 99%, Reagent Grade, Honeywell™

Product Code. 15647820 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
20 L
Packaging:
Glass bottle
Stainless steel can
Unit Size:
1 L
100mL
2.5 L
20 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15647820 20 L Stainless steel can 20 L
15617820 100 mL Glass bottle 100mL
15627820 1 L Glass bottle 1 L
15637820 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15647820 Supplier Honeywell Research Chemicals Supplier No. D5680220L

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-50-1
Molecular Formula C6H4Cl2
Molecular Weight (g/mol) 146.998
MDL Number MFCD00000535
InChI Key RFFLAFLAYFXFSW-UHFFFAOYSA-N
Synonym o-dichlorobenzene, chloroben, o-dichlorbenzol, dilantin db, ortho-dichlorobenzene, 2-dichlorobenzene, o-dichlorobenzol, odcb, benzene, 1,2-dichloro, cloroben
PubChem CID 7239
ChEBI CHEBI:35290
IUPAC Name 1,2-dichlorobenzene
SMILES C1=CC=C(C(=C1)Cl)Cl

Specifications

Melting Point -17°C
Color Colorless to Yellow
Density 1.306g/mL at 25°C
Boiling Point 180°C (1013 hPa)
Explosion Limit 0.092
Flash Point 66°C (150.8°F)
Ignition Point 1198°F
Odor Aromatic
Packaging Stainless steel can
Quantity 20 L
UN Number UN1591
Beilstein 606078
Vapor Density 5.1 (vs air)
Formula Weight 147g/mol
Vapor Pressure 1.6mmHg (35°C), 1.2mmHg (20°C)
Percent Purity 0.99
Chemical Name or Material 1,2-Dichlorobenzene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.