missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichlorobenzene, Spectrophotometric Grade, 98%

Product Code. 11338337
Change view
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
Unit Size:
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11338337 4 L 4 L
11348337 1 L 1 L
11328337 4 x 1 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11338337 Supplier Thermo Scientific Alfa Aesar Supplier No. 032154.K7
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Maximum level of impurities: Evaporation residue 5ppm, H2O 0.02% | CAS: 95-50-1 | C6H4Cl2 | 146.998 g/mol

1,2-Dichlorobenzene is used in spectrophotometry and environmental testing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-50-1
Molecular Formula C6H4Cl2
Molecular Weight (g/mol) 146.998
MDL Number MFCD00000535
InChI Key RFFLAFLAYFXFSW-UHFFFAOYSA-N
Synonym o-dichlorobenzene, chloroben, o-dichlorbenzol, dilantin db, ortho-dichlorobenzene, 2-dichlorobenzene, o-dichlorobenzol, odcb, benzene, 1,2-dichloro, cloroben
PubChem CID 7239
ChEBI CHEBI:35290
IUPAC Name 1,2-dichlorobenzene
SMILES C1=CC=C(C(=C1)Cl)Cl

Specifications

Melting Point °C to 18°C
Color Colorless
Density 1.306 g/cm3 at 20°C
Boiling Point 180°C
Flash Point 68°C (154°F)
Ignition Point 640°C (1184°F)
Assay Percent Range 98%
Odor Pungent
Refractive Index 1.551
Quantity 4 L
Beilstein 606078
Solubility Information Miscible with water and most of the organic solvents.
Formula Weight 147
Vapor Pressure 1.33 hPa (1mm Hg) at 20°C
Grade Spectrophotometric
Concentration or Composition (by Analyte or Components) 1, 2-Dichlorobenzene
Physical Form Liquid
Chemical Name or Material 1, 2-Dichlorobenzene, Filtered through 0.2μ filters.
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.