missing translation for 'onlineSavingsMsg'
Learn More

1,2-dichloro-4-iodobenzene, Thermo Scientific™

Product Code. 10456774
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10456774 100 g 100g
10779383 10 g 10g
10204624 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10456774 Supplier Thermo Scientific Maybridge Supplier No. BTB06155FA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 20555-91-3
Molecular Formula C6H3Cl2I
Molecular Weight (g/mol) 272.894
InChI Key NADPFZNWCQIJJW-UHFFFAOYSA-N
Synonym 3,4-dichloroiodobenzene, benzene, 1,2-dichloro-4-iodo, maybridge1_001437, pubchem3691, acmc-209fbv, 3,4-dichloro-iodobenzene, 1,2-dichloro-5-iodobenzene, 1-iodo-3,4-dichlorobenzene, 3,4-dichloro-1-iodobenzene, 4-iodo-1,2-dichlorobenzene
PubChem CID 88591
IUPAC Name 1,2-dichloro-4-iodobenzene
SMILES C1=CC(=C(C=C1I)Cl)Cl

Specifications

Assay Percent Range 97%
Packaging Amber glass bottle
Linear Formula IC6H3Cl2
Quantity 100 g
Formula Weight 272.9
Percent Purity 97%
Chemical Name or Material 1, 2-Dichloro-4-iodobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.