missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichloro-4-fluorobenzene, 98+%

Product Code. 11480153
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11480153 10 g 10g
11490153 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11480153 Supplier Thermo Scientific Alfa Aesar Supplier No. A11357.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1435-49-0 | C6H3Cl2F | 164.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1435-49-0
Molecular Formula C6H3Cl2F
Molecular Weight (g/mol) 164.99
MDL Number MFCD00018119
InChI Key QSDKXMVGRLVIQV-UHFFFAOYSA-N
Synonym 3,4-dichlorofluorobenzene, 3,4-dichloro-1-fluorobenzene, benzene, 1,2-dichloro-4-fluoro, pubchem3453, 3,4 dichlorofluorobenzene, acmc-1byo2, ksc494g9b, 1,2-dichloro-4-fluoro-benzene
PubChem CID 74028
IUPAC Name 1,2-dichloro-4-fluorobenzene
SMILES FC1=CC=C(Cl)C(Cl)=C1

Specifications

Melting Point -1°C
Density 1.403
Boiling Point 168°C to 171°C
Flash Point 64°C (147°F)
Refractive Index 1.524
Quantity 10 g
Beilstein 1861278
Formula Weight 165
Percent Purity ≥98%
Chemical Name or Material 1,2-Dichloro-4-fluorobenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.