missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichloro-3-iodobenzene, 98+%, Thermo Scientific™

Product Code. 11404763
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11404763 10 g 10g
11414763 50 g 50g
11424763 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11404763 Supplier Thermo Scientific Alfa Aesar Supplier No. A11647.09

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2401-21-0
Molecular Formula C6H3Cl2I
Molecular Weight (g/mol) 272.894
MDL Number MFCD00001034
InChI Key VGJKBWPZBVBXGI-UHFFFAOYSA-N
Synonym 2,3-dichloroiodobenzene, 2,3-dichloro-1-iodobenzene, 2,3-dichloro iodobenzene, dichloroiodobenzene, benzene, dichloroiodo, benzene, 1,2-dichloro-3-iodo, 1,2-dichloro-3-iodo-benzene, pubchem3690, 2,3-dichloro-iodobenzene, acmc-209g7t
PubChem CID 75459
IUPAC Name 1,2-dichloro-3-iodobenzene
SMILES C1=CC(=C(C(=C1)I)Cl)Cl

Specifications

Melting Point 33°C to 37°C
Boiling Point 121°C to 123°C (16 mmHg)
Flash Point >110°C (230°F)
Quantity 10 g
Beilstein 2247168
Sensitivity Light sensitive
Formula Weight 272.9
Percent Purity ≥98%
Chemical Name or Material 1,2-Dichloro-3-iodobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.