missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromotetrafluorobenzene, 99%

Product Code. 10076393 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10076393 5 g Glass bottle 5g
10115850 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10076393 Supplier Thermo Scientific Acros Supplier No. 311350050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 827-08-7 | C6Br2F4 | 307.88 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 827-08-7
Molecular Formula C6Br2F4
Molecular Weight (g/mol) 307.88
MDL Number MFCD00039209
InChI Key IPLUWQPTPKNBRD-UHFFFAOYSA-N
Synonym 1,2-dibromotetrafluorobenzene, 3,4,5,6-tetrafluoro-1,2-dibromobenzene, tetrafluoro-1,2-dibromobenzene, benzene, 1,2-dibromotetrafluoro, benzene, 1,2-dibromo-3,4,5,6-tetrafluoro, 1,2-dibromoperfluorobenzene, dtfb, pubchem2195, dibromotetrafluorobenzene, acmc-20ap6i
PubChem CID 70007
IUPAC Name 1,2-dibromo-3,4,5,6-tetrafluorobenzene
SMILES C1(=C(C(=C(C(=C1F)Br)Br)F)F)F

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 2.2300g/mL
Boiling Point 198.0°C
Flash Point 98°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula Br2C6F4
Refractive Index 1.5150 to 1.5170
Quantity 5 g
Specific Gravity 2.23
Formula Weight 307.88
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1, 2-Dibromotetrafluorobenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.