missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromotetrafluorobenzene, 99%, Thermo Scientific Chemicals

Catalog Number 10115850
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
827-08-7
C6Br2F4
307.88
MFCD00039209
IPLUWQPTPKNBRD-UHFFFAOYSA-N
1,2-dibromotetrafluorobenzene, 3,4,5,6-tetrafluoro-1,2-dibromobenzene, tetrafluoro-1,2-dibromobenzene, benzene, 1,2-dibromotetrafluoro, benzene, 1,2-dibromo-3,4,5,6-tetrafluoro, 1,2-dibromoperfluorobenzene, dtfb, pubchem2195, dibromotetrafluorobenzene, acmc-20ap6i
70007
1,2-dibromo-3,4,5,6-tetrafluorobenzene
C1(=C(C(=C(C(=C1F)Br)Br)F)F)F
This item is not returnable. View return policy
827-08-7
C6Br2F4
307.88
MFCD00039209
IPLUWQPTPKNBRD-UHFFFAOYSA-N
1,2-dibromotetrafluorobenzene, 3,4,5,6-tetrafluoro-1,2-dibromobenzene, tetrafluoro-1,2-dibromobenzene, benzene, 1,2-dibromotetrafluoro, benzene, 1,2-dibromo-3,4,5,6-tetrafluoro, 1,2-dibromoperfluorobenzene, dtfb, pubchem2195, dibromotetrafluorobenzene, acmc-20ap6i
70007
1,2-dibromo-3,4,5,6-tetrafluorobenzene
C1(=C(C(=C(C(=C1F)Br)Br)F)F)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 2.2300g/mL
Boiling Point 198.0°C
Flash Point 98°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula Br2C6F4
Refractive Index 1.5150 to 1.5170
Quantity 1 g
Specific Gravity 2.23
Formula Weight 307.88
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1, 2-Dibromotetrafluorobenzene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'