missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Dibromotetrachloroethane, TRC
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
Specifications
Specifications
| Chemical Name or Material | 1,2-Dibromotetrachloroethane |
| CAS | 630-25-1 |
| Molecular Formula | C2Br2Cl4 |
| Quantity | 5 g |
| Synonym | 1,2-Dibromo-1,1,2,2-tetrachloroethane, DBTCE, sym-Dibromotetrachloroethane |
| InChI Formula | InChI=1S/C2Br2Cl4/c3-1(5,6)2(4,7)8 |
| SMILES | ClC(Cl)(Br)C(Cl)(Cl)Br |
| IUPAC Name | 1,2-dibromo-1,1,2,2-tetrachloroethane |
| Molecular Weight (g/mol) | 325.64 |
| Formula Weight | 321.7121 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction