missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromoethane-d{4}, 99%(Isotopic)

Product Code. 11410860
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410860 5 g 5g
11420860 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11410860 Supplier Thermo Scientific Alfa Aesar Supplier No. 042279.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 22581-63-1 | C2H4Br2 | 191.886 g/mol

It is employed as a mutagenesis research chemical. Dibromoethane was shown to cause DNA single-strand breaks in isolated rat hepatocytes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22581-63-1
Molecular Formula C2H4Br2
Molecular Weight (g/mol) 191.886
MDL Number MFCD00037499
InChI Key PAAZPARNPHGIKF-LNLMKGTHSA-N
Synonym 1,2-dibromoethane-d4, 1,2-dibromo-1,1,2,2-2h4 ethane, dibromo 2 h? ethane, dibromoethane-d4, ethane-d4, 1,2-dibromo, ethylene-d4 dibromide, 2h4-edb, tetradeutero-1,2-dibromoethane, 1,2-dibromoethane-d4, 99 atom % d, 1,2-dibromoethane-d4 99.0 atom % d
PubChem CID 123404
IUPAC Name 1,2-dibromo-1,1,2,2-tetradeuterioethane
SMILES C(CBr)Br

Specifications

Physical Form Liquid
Density 2.20 g/mL
Odor Slightly Sweetish
Refractive Index 1.536
Quantity 5 g
Melting Point 9°C
Boiling Point 132°C
UN Number UN1605
Solubility Information Insoluble in water. Soluble in chloroform.
Formula Weight 191.89
Percent Purity 99%
Chemical Name or Material 1,2-Dibromoethane-d4, Isotopic
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.