missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromoethane, 99%

Product Code. 10346270 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic Bottle
Plastic Drum
Unit Size:
1 L
10 L
2.5 L
25mL
500mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10346270 1 L Glass bottle 1 L
10695142 25 mL Glass bottle 25mL
10244520 500 mL Glass bottle 500mL
11974481 2.5 L Plastic Bottle 2.5 L
15199153 10 L Plastic Drum 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Product Code. 10346270 Supplier Thermo Scientific Supplier No. 112790010
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

CAS: 106-93-4 | C2H4Br2 | 187.86 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-93-4
Molecular Formula C2H4Br2
Molecular Weight (g/mol) 187.86
InChI Key PAAZPARNPHGIKF-UHFFFAOYSA-N
Synonym ethylene dibromide, ethylene bromide, sym-dibromoethane, ethane, 1,2-dibromo, alpha,beta-dibromoethane, bromuro di etile, 1,2-dibromaethan, 1,2-dibroomethaan, 1,2-ethylene dibromide, aadibroom
PubChem CID 7839
ChEBI CHEBI:28534
IUPAC Name 1,2-dibromoethane
SMILES C(CBr)Br

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 9.0°C to 10.0°C
Color Colorless to Yellow
Density 2.1730g/mL
Boiling Point 131.0°C to 132.0°C
Flash Point >104°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula BrCH2CH2Br
Refractive Index 1.5375 to 1.5395
Quantity 1 L
Beilstein 01,9
Fieser 01,374; 04,126; 05,187; 12,570
Merck Index 15,385
Specific Gravity 2.173
Solubility Information Solubility in water: 4g/L (20°C). Other solubilities: miscible with most organic solvents
Formula Weight 187.86
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1,2-Dibromoethane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.