missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromoethane, 98%

Product Code. 15493057
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15493057 250 g 250g
15473057 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15493057 Supplier Thermo Scientific Alfa Aesar Supplier No. A12766.30
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 106-93-4 | C2H4Br2 | 187.862 g/mol

1,2-dibromoethane is widely used as a bromine source in organic synthesis. It is utilized as a fumigant for treatment of logs for termites and beetles, for control of moths in beehives. It finds the application to make vinyl bromide, a precursor to some fire retardants. It is used as an intermediate in the preparation of dyes and pharmaceuticals. It is also used as a lead scavenger in antiknock mixtures added to gasolines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-dibromoethane is widely used as a bromine source in organic synthesis. It is utilized as a fumigant for treatment of logs for termites and beetles, for control of moths in beehives. It finds the application to make vinyl bromide, a precursor to some fire retardants. It is used as an intermediate in the preparation of dyes and pharmaceuticals. It is also used as a lead scavenger in antiknock mixtures added to gasolines.

Solubility
Miscible with alcohol, acetone, benzene, diethyl ether and ethyl acetate. Slightly miscible with water.

Notes
Special handling precautions required. View MSDS prior to purchase. MSDS are available online at www.alfa.com.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-93-4
Molecular Formula C2H4Br2
Molecular Weight (g/mol) 187.862
MDL Number MFCD00000233
InChI Key PAAZPARNPHGIKF-UHFFFAOYSA-N
Synonym ethylene dibromide, ethylene bromide, sym-dibromoethane, ethane, 1,2-dibromo, alpha,beta-dibromoethane, bromuro di etile, 1,2-dibromaethan, 1,2-dibroomethaan, 1,2-ethylene dibromide, aadibroom
PubChem CID 7839
ChEBI CHEBI:28534
IUPAC Name 1,2-dibromoethane
SMILES C(CBr)Br

Specifications

Melting Point 9°C to 10°C
Density 2.179
Boiling Point 132°C
Flash Point None
Linear Formula BrCH2CH2Br
Refractive Index 1.5385
Quantity 250 g
UN Number UN1605
Sensitivity Light sensitive
Merck Index 14,3796
Solubility Information Miscible with alcohol,acetone,benzene,diethyl ether and ethyl acetate. Slightly miscible with water.
Formula Weight 187.87
Percent Purity 99%
Chemical Name or Material 1,2-Dibromoethane
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1,2-Dibromoethane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H350-May cause cancer.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P330-Rinse mouth.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Ethylene dibromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.