missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Dibromoethane, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 mL
500 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic Bottle
Plastic Drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Melting Point | 9.0°C to 10.0°C |
Color | Colorless to Yellow |
Density | 2.1730g/mL |
Boiling Point | 131.0°C to 132.0°C |
Flash Point | >104°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (GC) |
Packaging | Glass bottle |
Show More |
Safety and Handling
Product Identifier
- 1,2-Dibromoethane
- Danger
- Acute toxicity Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Carcinogenicity Category 1B
- LONG-TERM AQUATIC HAZARD Chronic 2
- H319-Causes serious eye irritation.
- H315-Causes skin irritation.
- H350-May cause cancer.
- H411-Toxic to aquatic life with long lasting effects.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
- H301-Toxic if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'