missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromocyclopentene, 97%

Product Code. 10410514 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10410514 5 g Glass bottle 5g
10336824 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10410514 Supplier Thermo Scientific Acros Supplier No. 399420050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 75415-78-0 | C5H6Br2 | 225.9 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75415-78-0
Molecular Formula C5H6Br2
Molecular Weight (g/mol) 225.9
InChI Key PNWFXPGGROADNS-UHFFFAOYSA-N
Synonym 1,2-dibromocyclopent-1-ene, dibromocyclopentene, pubchem13714, acmc-20ap6k, 1,2-bis bromanyl cyclopentene, 1,2-dibromocyclopentene
PubChem CID 4438528
IUPAC Name 1,2-dibromocyclopentene
SMILES C1CC(=C(C1)Br)Br

Specifications

Density 1.8950g/mL
Boiling Point 78.0°C (5.0 mmHg)
Flash Point >110°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Specific Gravity 1.895
Formula Weight 225.9
Percent Purity 97%
Chemical Name or Material 1, 2-Dibromocyclopentene
compliance-icons
Product Identifier
  • 1,2-Dibromocyclopentene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.