missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromobenzene, TRC

Product Code. 30380753
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30380753 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30380753 Supplier Toronto Research Chemicals Supplier No. TRCD4242905G
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2-Dibromobenzene
CAS 583-53-9
Molecular Formula C6 H4 Br2
Quantity 5 g
Synonym Benzene, o-dibromo- (8CI), 1,2-Dibromobenzene, NSC 60643, o-Dibromobenzene
InChI Formula InChI=1S/C6H4Br2/c7-5-3-1-2-4-6(5)8/h1-4H
SMILES Brc1ccccc1Br
IUPAC Name 1,2-dibromobenzene
Molecular Weight (g/mol) 235.9
Formula Weight 233.868
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.