Learn More
1,2-Dibromobenzene, 98%
Description
1,2-Dibromobenzene is used as a precursor in the preparation of 3-(2-bromophenylthio)-4H-[1]benzopyran-4-one using copper as a reagent. It is involved in the preparation of isoindoloquinazolinones by the carbonylation reaction with 2-aminobenzyl amine using palladium as the catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1,2-Dibromobenzene is used as a precursor in the preparation of 3-(2-bromophenylthio)-4H-[1]benzopyran-4-one using copper as a reagent. It is involved in the preparation of isoindoloquinazolinones by the carbonylation reaction with 2-aminobenzyl amine using palladium as the catalyst.
Solubility
Miscible with ethanol, ethyl ether, acetone, chloroform, acetic acid, benzene, petroleum ether and tetrachloromethane. Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 4°C to 6°C |
| Density | 1.985 |
| Boiling Point | 224°C to 226°C |
| Flash Point | 91°C (195°F) |
| Odor | Strong |
| Refractive Index | 1.611 |
| Quantity | 250 g |
| UN Number | UN3082 |
| Beilstein | 970241 |
| Solubility Information | Miscible with ethanol,ethyl ether,acetone,chloroform,acetic acid,benzene,petroleum ether and tetrachloromethane. Immiscible with water. |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.