missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromobenzene, 98%

Product Code. 11457826
Click to view available options
Quantity:
25 g
50 g
100 g
250 g
Unit Size:
100g
250g
25g
50g
This item is not returnable. View return policy

Product Code. 11457826

Brand: Thermo Scientific Alfa Aesar A14765.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 583-53-9 | C6H4Br2 | 235.906 g/mol

1,2-Dibromobenzene is used as a precursor in the preparation of 3-(2-bromophenylthio)-4H-[1]benzopyran-4-one using copper as a reagent. It is involved in the preparation of isoindoloquinazolinones by the carbonylation reaction with 2-aminobenzyl amine using palladium as the catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Dibromobenzene is used as a precursor in the preparation of 3-(2-bromophenylthio)-4H-[1]benzopyran-4-one using copper as a reagent. It is involved in the preparation of isoindoloquinazolinones by the carbonylation reaction with 2-aminobenzyl amine using palladium as the catalyst.

Solubility
Miscible with ethanol, ethyl ether, acetone, chloroform, acetic acid, benzene, petroleum ether and tetrachloromethane. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 583-53-9
Molecular Formula C6H4Br2
Molecular Weight (g/mol) 235.906
MDL Number MFCD00000057
InChI Key WQONPSCCEXUXTQ-UHFFFAOYSA-N
Synonym o-dibromobenzene, dibromobenzene, benzene, 1,2-dibromo, ortho-dibromobenzene, benzene, o-dibromo, benzene, dibromo, unii-52k9w7u6eh, dibromobenzene mixed isomers, 1,2 dibromobenzene, 1,6-dibromobenzene
PubChem CID 11414
ChEBI CHEBI:37152
IUPAC Name 1,2-dibromobenzene
SMILES C1=CC=C(C(=C1)Br)Br

Specifications

Melting Point 4°C to 6°C
Density 1.985
Boiling Point 224°C to 226°C
Flash Point 91°C (195°F)
Odor Strong
Refractive Index 1.611
Quantity 50 g
UN Number UN3082
Beilstein 970241
Solubility Information Miscible with ethanol,ethyl ether,acetone,chloroform,acetic acid,benzene,petroleum ether and tetrachloromethane. Immiscible with water.
Formula Weight 235.91
Percent Purity 98%
Chemical Name or Material 1,2-Dibromobenzene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,2-Dibromobenzene
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 1,2-Dibromobenzene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.