missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromobenzene, 98%

Product Code. 11454240
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
250 g
Unit Size:
100g
250g
25g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11454240 100 g 100g
11444240 25 g 25g
11457826 50 g 50g
11467826 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11454240 Supplier Thermo Scientific Alfa Aesar Supplier No. A14765.22
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 583-53-9 | C6H4Br2 | 235.906 g/mol

1,2-Dibromobenzene is used as a precursor in the preparation of 3-(2-bromophenylthio)-4H-[1]benzopyran-4-one using copper as a reagent. It is involved in the preparation of isoindoloquinazolinones by the carbonylation reaction with 2-aminobenzyl amine using palladium as the catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Dibromobenzene is used as a precursor in the preparation of 3-(2-bromophenylthio)-4H-[1]benzopyran-4-one using copper as a reagent. It is involved in the preparation of isoindoloquinazolinones by the carbonylation reaction with 2-aminobenzyl amine using palladium as the catalyst.

Solubility
Miscible with ethanol, ethyl ether, acetone, chloroform, acetic acid, benzene, petroleum ether and tetrachloromethane. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 583-53-9
Molecular Formula C6H4Br2
Molecular Weight (g/mol) 235.906
MDL Number MFCD00000057
InChI Key WQONPSCCEXUXTQ-UHFFFAOYSA-N
Synonym o-dibromobenzene, dibromobenzene, benzene, 1,2-dibromo, ortho-dibromobenzene, benzene, o-dibromo, benzene, dibromo, unii-52k9w7u6eh, dibromobenzene mixed isomers, 1,2 dibromobenzene, 1,6-dibromobenzene
PubChem CID 11414
ChEBI CHEBI:37152
IUPAC Name 1,2-dibromobenzene
SMILES C1=CC=C(C(=C1)Br)Br

Specifikationer

Melting Point 4°C to 6°C
Density 1.985
Boiling Point 224°C to 226°C
Flash Point 91°C (195°F)
Odor Strong
Refractive Index 1.611
Quantity 100 g
UN Number UN3082
Beilstein 970241
Solubility Information Miscible with ethanol,ethyl ether,acetone,chloroform,acetic acid,benzene,petroleum ether and tetrachloromethane. Immiscible with water.
Formula Weight 235.91
Percent Purity 98%
Chemical Name or Material 1,2-Dibromobenzene
Visa mer Visa mindre

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.