missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromo-4,5-(methylenedioxy)benzene, 98%, Thermo Scientific™

Catalog Number p-7058393
Click to view available options
Quantity:
50 g
This item is not returnable. View return policy
5279-32-3
C7H4Br2O2
279.92
MFCD00094476
WPYAICCSYGUFTK-UHFFFAOYSA-N
5,6-dibromobenzo d 1,3 dioxole, 1,2-dibromo-4,5-methylenedioxy benzene, 5,6-dibromo-2h-1,3-benzodioxole, 1,2-dibromo-4,5-methylenedioxybenzene, acmc-20ap6l, 1,3-benzodioxole,5,6-dibromo, 5,6-dibromo-benzo-1,3 dioxole, 5,6-bis bromanyl-1,3-benzodioxole, 4,5-dibromo-1,2-methylenedioxybenzene, 4,5-methylenedioxy-1,2-dibromobenzene
225814
BrC1=CC2=C(OCO2)C=C1Br
This item is not returnable. View return policy
5279-32-3
C7H4Br2O2
279.92
MFCD00094476
WPYAICCSYGUFTK-UHFFFAOYSA-N
5,6-dibromobenzo d 1,3 dioxole, 1,2-dibromo-4,5-methylenedioxy benzene, 5,6-dibromo-2h-1,3-benzodioxole, 1,2-dibromo-4,5-methylenedioxybenzene, acmc-20ap6l, 1,3-benzodioxole,5,6-dibromo, 5,6-dibromo-benzo-1,3 dioxole, 5,6-bis bromanyl-1,3-benzodioxole, 4,5-dibromo-1,2-methylenedioxybenzene, 4,5-methylenedioxy-1,2-dibromobenzene
225814
BrC1=CC2=C(OCO2)C=C1Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 83°C to 85°C
Quantity 50 g
Beilstein 145633
IUPAC Name 5,6-dibromo-1,3-benzodioxole
Formula Weight 279.93
Percent Purity 98%
Chemical Name or Material 1,2-Dibromo-4,5-(methylenedioxy)benzene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'