missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromo-4,5-difluorobenzene, 98%

Product Code. 11472547
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11472547 5 g 5g
11482547 25 g 25g
11492547 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11472547 Supplier Thermo Scientific Alfa Aesar Supplier No. A14804.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 64695-78-9 | C6H2Br2F2 | 271.887 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64695-78-9
Molecular Formula C6H2Br2F2
Molecular Weight (g/mol) 271.887
MDL Number MFCD00009890
InChI Key JTEZQWOKRHOKDG-UHFFFAOYSA-N
Synonym benzene, 1,2-dibromo-4,5-difluoro, 1,2-dibromo-4,5-difluoro-benzene, 1,2-difluoro-4,5-dibromobenzene, zlchem 536, pubchem3440, acmc-1biw8, 1,2-dibromo-4,5-difluorobenzene, benzene, 1,2-dibromo-4,5-difluoro-;1,2-dibromo-4,5-difl
PubChem CID 94949
IUPAC Name 1,2-dibromo-4,5-difluorobenzene
SMILES C1=C(C(=CC(=C1Br)Br)F)F

Specifications

Melting Point 31°C to 35°C
Quantity 5 g
Beilstein 1941132
Formula Weight 271.89
Percent Purity 98%
Chemical Name or Material 1,2-Dibromo-4,5-difluorobenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.