missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromo-2-methylpropane, 98%

Product Code. 10156790 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
Packaging:
Glass bottle
Unit Size:
250g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10156790 5 g Glass bottle 5g
10647181 25 g Glass bottle 25g
10240303 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10156790 Supplier Thermo Scientific Acros Supplier No. 406490050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 594-34-3 | C4H8Br2 | 215.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 594-34-3
Molecular Formula C4H8Br2
Molecular Weight (g/mol) 215.92
MDL Number MFCD00000126
InChI Key SDTXSEXYPROZSZ-UHFFFAOYSA-N
Synonym isobutylene bromide, 1,2-dibromoisobutane, acmc-1aolc, propane,2-dibromo-2-methyl, propane,1,2-dibromo-2-methyl, 1,2-dibromo-2-methylpropane
PubChem CID 222657
IUPAC Name 1,2-dibromo-2-methylpropane
SMILES CC(C)(CBr)Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 1.7900g/mL
Boiling Point 150.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2CBrCH2Br
Refractive Index 1.5083 to 1.5103
Quantity 5 g
Specific Gravity 1.79
Solubility Information Solubility in water: slightly soluble
Formula Weight 215.92
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1, 2-Dibromo-2-methylpropane, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,2-Dibromo-2-methylpropane
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H351-Suspected of causing cancer.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.