missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromo-1-iodotrifluoroethane, 97%, Thermo Scientific™

Product Code. 11447104
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11447104 5 g 5g
11437104 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11447104 Supplier Thermo Scientific Alfa Aesar Supplier No. B21532.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 216394-01-3
Molecular Formula C2Br2F3I
Molecular Weight (g/mol) 367.73
MDL Number MFCD00042521
InChI Key YUBJHLYGOOZDRN-UHFFFAOYNA-N
Synonym 1,2-dibromo-1-iodotrifluoroethane, 1,2-dibromo-2-iodotrifluoroethane, 1,2-dibromo-1,2,2-trifluoro-1-iodoethane, ethane,1,2-dibromo-1,1,2-trifluoro-2-iodo
PubChem CID 2736780
IUPAC Name 1,2-dibromo-1,1,2-trifluoro-2-iodoethane
SMILES FC(F)(Br)C(F)(Br)I

Spezifikation

Density 1.515
Boiling Point 63°C to 65°C (32 mmHg)
Linear Formula BrCFICF2Br
Quantity 5 g
Formula Weight 367.73
Percent Purity 97%
Chemical Name or Material 1,2-Dibromo-1-iodotrifluoroethane
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.