missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromo-1,1,2-trichloroethane, 99%, Thermo Scientific™

Product Code. 10323432 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10323432 5 g Glass bottle 5g
10477382 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10323432 Supplier Thermo Scientific Acros Supplier No. 279030050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 13749-38-7
Molecular Formula C2HBr2Cl3
Molecular Weight (g/mol) 291.2
MDL Number MFCD00018841
InChI Key VXKOVUFOVCJHBA-UHFFFAOYSA-N
Synonym ethane,1,2-dibromo-1,1,2-trichloro, acmc-20ap6p, 1,2-dibromotrichloro ethane
PubChem CID 114562
IUPAC Name 1,2-dibromo-1,1,2-trichloroethane
SMILES C(C(Cl)(Cl)Br)(Cl)Br

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Flash Point >200°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5700 to 1.5720
Quantity 5 g
Formula Weight 291.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1,2-Dibromo-1,1,2-trichloroethane, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.