missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibenzoylethane, 98+%

Product Code. 11362636
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11362636 5 g 5g
11352636 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11362636 Supplier Thermo Scientific Alfa Aesar Supplier No. L08652.06
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 495-71-6 | C16H14O2 | 238.286 g/mol

1,2-Dibenzoylethane is effective in inhibiting the mammary DMBA-DNA adduct formation. This inhibitory effect on mammary DNA adduct formation was associated with increased liver activities of glutathione S-transferase, QR, and 7-ethoxyresorufin-O-deethylase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Dibenzoylethane is effective in inhibiting the mammary DMBA-DNA adduct formation. This inhibitory effect on mammary DNA adduct formation was associated with increased liver activities of glutathione S-transferase, QR, and 7-ethoxyresorufin-O-deethylase.

Solubility
Soluble in acetone. Insoluble in water.

Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 495-71-6
Molecular Formula C16H14O2
Molecular Weight (g/mol) 238.286
MDL Number MFCD00037818
InChI Key OSWWFLDIIGGSJV-UHFFFAOYSA-N
Synonym 1,2-dibenzoylethane, 1,4-diphenyl-1,4-butanedione, 1,4-butanedione, 1,4-diphenyl, biphenacyl, diphenacyl, 2,2-biacetophenone, acmc-1bd9f, ethane, 1,2-dibenzoyl, succinophenone diphenacyl, cambridge id 5102527
PubChem CID 136322
IUPAC Name 1,4-diphenylbutane-1,4-dione
SMILES C1=CC=C(C=C1)C(=O)CCC(=O)C2=CC=CC=C2

Specifications

Melting Point 144°C to 148°C
Boiling Point 260°C (15 mmHg)
Quantity 5 g
Beilstein 782937
Solubility Information Soluble in acetone. Insoluble in water.
Formula Weight 238.29
Percent Purity ≥98%
Chemical Name or Material 1,2-Dibenzoylethane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.